Learn More
Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder
The touch-screen reader has 8GB on board memory to provide virtually unlimited data logging capacity
Brand:  Digi-Sense™ 20250-49
 View more versions of this product
Product Code. 15840016
  • 11.15 € / Each
Estimated Shipment: 15-10-2025
Explore more special offers
Add to basket
Description
Description
  • Includes A2LA accredited NIST-Traceable Calibration Certificate
  • Measures, logs, and displays temperature, humidity, and dew point
  • Removable sensor for local or remote measurements using extension cable included
  • Wall mount or desk mount operation
  • Resident self-booting software, no installation necessary
  • Comes with NIST calibration
  • Stored data downloads to computer via USB cable provided in CSV format
Services
2 Years
Specifications
Specifications
Replacement USB Cable
Temperature/RH Touch Screen Recorder with NIST Traceable Calibration (20250-40)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Digi-Sense™ Accessories for the 20250-40 Digi-Sense™ Temperature Humidity Touch-screen Recorder > Replacement USB Cable for 13245300 (20250-40)