Learn More
Thermo Scientific™ Accessories for the LP Vortex Mixer
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
Brand:  Thermo Scientific™ 88880121
 View more versions of this product
Product Code. 15288874
  • 16.48 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.
  •  Warranty: 2 years
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube Holder
12 mm
Tube Holder with Insert Retainer (dia. 12mm)
Vortex Mixer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.