Learn More
Fisher Scientific™ Accu500 Water System
Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.
Brand:  Fisher Scientific™ RD0P020UEK
 View more versions of this product
Product Code. 15459699
  • 9895.00 € / Each
Estimated Shipment: 01-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
<20μS/cm
Tap Water
Accu500 UV Ultrapure Water System
Accu500
1.5 L/min.
5°C to 45°C
48 cm
<0.001 EU/mL with a terminal ultrafiltration cartridge
18.2 Mohm.cm
1 to 6 bar
<1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
20 L/hr.
30 cm
51 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisher Scientific™ Accu500 Water System >