missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AE series ATC temperature probe

Fisher Scientific™ accumet™ AE series ATC temperature probes support the accumet AE150 pH meter for routine pH measurements at a budget-friendly price.

Brand:  Fisherbrand™ 13-620-21

40.79 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15573633

  • 48.55 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Fisher Scientific™ accumet™ AE series ATC temperature probes support the accumet AE150 pH meter for routine pH measurements at a budget-friendly price.

Product Features:
  • ATC temperature probe
  • Durable stainless steel body
  • 90mm or 120mm body length
  • 2.5mm phono plug connection
  • 1 meter cable

Services

6 months

TRUSTED_SUSTAINABILITY
Specifications

Specifications

13-636-AE150 accumet AE150 pH Meter
0°C to 100°C
120 mm
0°C to 100°C
Temperature Probe
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers