missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AE6 Portable pH/ORP Meter

The AE6 Portable pH Meters are ideal for quick, accurate conductivity or TDS checks in laboratories, field and schools.

528.00 € - 769.00 €

Specifications

Accuracy (mV) ±1
Accuracy (pH) ±0.01
Accuracy (Temperature) ±0.5°C
For Use With (Application) Water Quality Testing
Calibration 1.68, 4.01, 7, 10.01, 12.45
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15523653
View accessory products View Documents
Fisherbrand™
13-636-AE6
AE6 pH/ORP Meter, Rubber Armor/Stand, 3-in-1 pH/Temperature Combination Electrode ECFC7352901B and pH Carrying Kit
528.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15533653
View accessory products View Documents
Fisherbrand™
13-636-AE6B
AE6 pH/ORP Meter, Rubber Armor/Stand, Double Junction pH Electrode ECFC7252201B and pH Carrying Kit
769.00 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
Description

Description

Fisherbrand accumet AE6 Portable pH Meter, Includes: pH/Temp electrode, Accuracy: +/- 0.01pH, +/-0.5 deg.C, +/- 1mV, Range: -1000 to 1000 mV, 0 to 14, Resolution: 0.01, 1mV, 0.1 deg.C, Battery Type: 4 x AAA

  • Multipoint calibration (up to 5 points) with auto-buffer recognition
  • Choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • For analyzing water in pools and spas, lithographic processes, boiler and cooling tower water analysis, environmental and agricultural purposes, and all types of quality assurance and water quality testing
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold function momentarily freezes reading for easy viewing and recording
  • Auto-power off saves battery power after 17 minutes of non-use
  • Self-diagnostic with message codes for easy troubleshooting
  • Large custom LCD provides optimum viewing, even at a distance
  • Rubber boot shields protect from accidental knocks and feature sturdy, built-in stand for easy benchtop operation
  • Optional rugged carrying case provides handy on-site meter calibration and measurement
Specifications

Specifications

±1
±0.5°C
1.68, 4.01, 7, 10.01, 12.45
0.01
1 mV
0°C to 100°C
Automatic/Manual (0°C to 100°C)
Portable Meter
±0.01
Water Quality Testing
0 to 14
-1000 to +1000 mV
1 to 5
0.1°C
AAA
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ accumet™ AE6 Portable pH/ORP Meter

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.