missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AE6 Portable pH/ORP Meter

The AE6 Portable pH Meters are ideal for quick, accurate conductivity or TDS checks in laboratories, field and schools.

Brand:  Fisherbrand™ 13-636-AE6B

 View more versions of this product

Product Code. 15533653

  • 769.00 â‚¬ / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket

Includes: Double junction pH electrode and ATC probe
This item is not returnable. View return policy

Description

Description

Fisherbrand accumet AE6 Portable pH Meter, Includes: pH/Temp electrode, Accuracy: +/- 0.01pH, +/-0.5 deg.C, +/- 1mV, Range: -1000 to 1000 mV, 0 to 14, Resolution: 0.01, 1mV, 0.1 deg.C, Battery Type: 4 x AAA

  • Multipoint calibration (up to 5 points) with auto-buffer recognition
  • Choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • For analyzing water in pools and spas, lithographic processes, boiler and cooling tower water analysis, environmental and agricultural purposes, and all types of quality assurance and water quality testing
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold function momentarily freezes reading for easy viewing and recording
  • Auto-power off saves battery power after 17 minutes of non-use
  • Self-diagnostic with message codes for easy troubleshooting
  • Large custom LCD provides optimum viewing, even at a distance
  • Rubber boot shields protect from accidental knocks and feature sturdy, built-in stand for easy benchtop operation
  • Optional rugged carrying case provides handy on-site meter calibration and measurement
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AE6 pH/ORP Meter, Rubber Armor/Stand, Double Junction pH Electrode ECFC7252201B and pH Carrying Kit
±1
Water Quality Testing
0 to 14
-1000 to +1000 mV
1 to 5
0.1°C
AAA
±0.5°C
±0.01
1.68, 4.01, 7, 10.01, 12.45
0.01
1 mV
0°C to 100°C
Automatic/Manual (0°C to 100°C)
Portable Meter
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand - accumet AE6 pH and AE6C conductivity meters
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand accumet AE6 pH and AE6C conductivity meters

    SDS
    Documents

    Documents

    Brochures

    Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Fisherbrand™ accumet™ AE6 Portable pH/ORP Meter > AE6 Portable pH Meter, Includes: AE6 pH/ORP Meter, Rubber Armor/Stand, Double Junction pH Electrode ECFC7252201B and pH Carrying Kit; Battery Type: AAA; Accuracy (mV): ±1; Range (mV): -1000 to +1000 mV

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.