Learn More
Fisherbrand™ accuQuatro U Pack Cartridge
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
Brand:  Fisherbrand™ RR100Q301
 View more versions of this product
Product Code. 15419719
  • 954.00 € / Each
Estimated Shipment: 08-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
Specifications
Accu100
AccuQuatro U pack Cartridge, Low TOC