Learn More
Fisherbrand™ accuQuatro U Pack Cartridge
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
892.00 € - 1050.00 €
Specifications
For Use With (Equipment) Accu100
View More Specs
Products 5
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15479709
View Documents
Fisherbrand™
RR100Q101
AccuQuatro U pack Cartridge, RO Feed
892.00 €
Each
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
15489709
View Documents
Fisherbrand™
RR100Q1KT
AccuQuatro U pack Cartridge Kit with 0.2μm final filter, RO feed
1050.00 €
Each
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
15499709
View Documents
Fisherbrand™
RR100Q201
AccuQuatro U pack Cartridge, DI Feed
892.00 €
Each
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
15409719
View Documents
Fisherbrand™
RR100Q2KT
AccuQuatro U pack Cartridge Kit with 0.2μm final filter, DI Feed
1050.00 €
Each
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
15419719
View Documents
Fisherbrand™
RR100Q301
AccuQuatro U pack Cartridge, Low TOC
954.00 €
Each
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
Description
Description
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
Specifications
Accu100