missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, glacial, ACS, 99.7+%, Thermo Scientific Chemicals

39.60 € - 167.84 €

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393358
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036289.AE
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11475160
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036289.AP
500 mL
45.53 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11485160
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036289.AY
4 x 500 mL
167.84 €
2000mL
Please to purchase this item. Need a web account? Register with us today!
13572310
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036289.K3
2 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11303368
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036289.K3
2 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

It is used in the production of cellulose acetate for photographic film, polyvinyl acetate for wood glue, synthetic fibers and fabrics. It is used as table vinegar, as preservative and as intermediate in the chemical industry. It is useful for the extraction of glazed ceramic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

64-19-7
118.1°C
C2H4O2
UN2789
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
ACS
40°C (104°F)
1.3721
16.6°C
99.7+%
MFCD00036152
14,55
Miscible with water.
CC(O)=O
60.05
CHEBI:15366
≥99.7%
1.049
Liquid
Acetic Acid, glacial, Glacial
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226

P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETIC ACID, GLACIAL

missing translation for 'einecsNumber' : 200-580-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only