missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss., 99-100%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Specifications
Specifications
Linear Formula | CH3CO2H |
UN Number | 2789 |
Beilstein | 506007 |
Formula Weight | 60.05g/mol |
Percent Purity | 99 to 100% |
Grade | puriss. |
Density | 1.049g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 16% (92°F), 4% (59°F) |
Flash Point | 40°C (104°F) |
Show More |
Safety and Handling


Product Identifier
- Acetic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetic acid, Puriss., 99-100%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction