missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 kg
30 kg
4 x 2.5 L
60 kg
6 x 1 L
200 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Linear Formula | CH3CO2H |
UN Number | 2789 |
Beilstein | 506007 |
Formula Weight | 60.05g/mol |
Percent Purity | 99.8 to 100.5% |
Grade | puriss. |
Density | 1.049 g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 16% (92°F), 4% (59°F) |
Flash Point | 40°C (104°F) |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
O3230 | Certificate of Analysis | 19/02/2025 | 27225-1L |
N1870 | Certificate of Analysis | 08/01/2025 | 27225-1L |
K3310 | Certificate of Analysis | 06/04/2023 | 27225-1L |
K2020 | Certificate of Analysis | 30/03/2023 | 27225-2.5L,27225-30KG,27225-4X2.5L,27225-60KG,27225-6X1L,27225-10KG,27225-1L |
Safety and Handling


Product Identifier
- Acetic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetic acid, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.8-100.5%, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction