missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss., 99-100%, Honeywell™
Puriss., 99-100%
58.39 € - 884.17 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15622890
|
Honeywell Research Chemicals
27221-1L |
1 L |
58.39 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15632890
|
Honeywell Research Chemicals
27221-2.5L |
2.5 L |
171.10 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15662890
|
Honeywell Research Chemicals
27221-5L |
5 L |
245.45 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15672890
|
Honeywell Research Chemicals
27221-6X1L |
6 x 1 L |
294.98 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15642890
|
Honeywell Research Chemicals
27221-4X2.5L |
4 x 2.5 L |
504.82 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15652890
|
Honeywell Research Chemicals
27221-4X5L |
4 x 5 L |
884.17 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
CH3CO2H | |
2789 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
60.05 | |
CHEBI:15366 | |
99 to 100% | |
1.049g/mL at 25°C | |
40°C (104°F) | |
2.07 (vs air) |
C2H4O2 | |
MFCD00036152 | |
506007 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
acetic acid | |
176 | |
60.05g/mol | |
puriss. | |
16% (92°F), 4% (59°F) | |
800°F | |
Acetic Acid |
Safety and Handling
P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
H226-H314
missing translation for 'einecsNumber' : 200-580-7
missing translation for 'rtecsNumber' : AF1225000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetic acid
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title