missing translation for 'onlineSavingsMsg'
Learn More
Sodium hypochlorite, 10-15% active chlorine, Thermo Scientific Chemicals Product Code.: 10401841

Sodium hypochlorite, 10-15% active chlorine, Thermo Scientific Chemicals

Product Code. 10401841
500 mL, Plastic bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Plastic bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10401841

Brand: Thermo Scientific Acros 219255000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9, 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solutionShow MoreShow Less
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Melting Point -16.0°C
Color Yellow
Physical Form Solution
Quantity 500 mL
Linear Formula NaOCl
Fieser 01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308
Merck Index 15,8762
Solubility Information Solubility in water: soluble
Formula Weight 74.44
Percent Purity ≥12%
Packaging Plastic bottle
Specific Gravity 1.2
Density 1.2000g/mL
Chemical Name or Material Sodium hypochlorite, 13% Active chlorine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462985 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number219250100,219255000,219250025
Lot NumberA0467172 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number219250100,219250250,219255000,219250025
Lot NumberA0462985 Certificate TypeCertificate of Analysis Date20/05/2024 Catalog Number219250100,219250250,219255000,219250025
Lot NumberA0460350 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number219250100,219250250,219255000,219250025
Lot NumberA0458700 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog Number219250100,219250250,219255000,219250025
compliance-icons compliance-icons
Product Identifier
  • Sodium hypochlorite
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hypochlorite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hypochlorite, 10-15% active chlorine, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.