missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid solution, 1.0M, Honeywell Fluka™
Volumetric, 1.0 M CH3COOH (1.0N)
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
PubChem CID | 176 |
ChEBI | CHEBI:15366 |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
64-19-7 | |
60.05 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
176 | |
acetic acid |
C2H4O2 | |
MFCD00036152 | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CHEBI:15366 | |
CC(O)=O |
Specifications
64-19-7 | |
CH3CO2H | |
NONH for all modes of transport | |
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
CC(O)=O | |
acetic acid | |
176 | |
60.05g/mol |
C2H4O2 | |
MFCD00036152 | |
506007 | |
QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
1.0M | |
60.05 | |
CHEBI:15366 | |
Acetic acid solution |
Safety and Handling
Product Identifier
- Acetic acid solution