missing translation for 'onlineSavingsMsg'
Learn More
Acetone, ACS, 99.5+%, Thermo Scientific Chemicals Product Code.: 11317328

Acetone, ACS, 99.5+%, Thermo Scientific Chemicals

Product Code. 11317328
10 mL
Click to view available options
Quantity:
10 mL
500 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy

Product Code. 11317328

Brand: Thermo Scientific Alfa Aesar 030698.AB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glasswares due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon, photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetoneShow MoreShow Less
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point °C to 94°C
Color Colorless
Appearance Colorless liquid
Density 0.791
Boiling Point 56°C
Quantity 10 mL
Flash Point -17°C (1°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.359
Assay Percent Range >99.5%
Vapor Pressure 247 hPa (185mm Hg) at 20°C
UN Number 1090; Colorless
Beilstein 635680
Solubility Information Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
Formula Weight 58.08
Grade ACS
Chemical Name or Material Acetone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN28L024 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number030698.K2,030698.M1,030698.AB,030698.AP,030698.K7
Lot NumberM30L007 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog Number030698.K2,030698.M1,030698.AB,030698.AP,030698.K7
Lot NumberY06K028 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog Number030698.K2,030698.M1,030698.AB,030698.AP,030698.K7
Lot NumberW18K038 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number030698.K2,030698.M1,030698.AB,030698.AP,030698.K7
Lot NumberU16K027 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number030698.K2,030698.M1,030698.AB,030698.AP,030698.K7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, ACS, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.