missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C

Brand:  Fisher Chemical A/0600/17

26.64 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.



 View more versions of this product

Product Code. 10162180

  • 124.00 € / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-64-1
2.5 L
0.32 mPaS at 20°C
C3H6O
Liquid
7
-95°C
CC(C)=O
58.08
CHEBI:15347
Amber Glass Bottle
247mbar at 20°C
2
MFCD00008765
Colorless
CSCPPACGZOOCGX-UHFFFAOYSA-N
56°C
propan-2-one
180
58.08g/mol
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527023 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberA/0600/17,A/0600/27,A/0600/PB17,A/0600/PC17,A/0600/PB08,A/0600/PC17X,A/0600/25X,A/0600/21,A/0600/15Z,A/0600/17Z,A/0600/15,A/0600/PC21,A/0600/JC21,A/0600/25SS,A/0600/25,A/0600/99SS
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™ > 2.5L, Amber glass bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.