missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS, 99.5+%, Thermo Scientific Chemicals

24.32 € - 146.33 €

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetoneShow MoreShow Less
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317328
SDS View Documents
Thermo Scientific Alfa Aesar
030698.AB
10 mL
24.32 €
10mL
Please to purchase this item. Need a web account? Register with us today!
11420520
SDS View Documents
Thermo Scientific Alfa Aesar
030698.AP
500 mL
27.80 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11307328
SDS View Documents
Thermo Scientific Alfa Aesar
030698.K2
1 L
39.40 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11327328
SDS View Documents
Thermo Scientific Alfa Aesar
030698.M1
4 x 1 L
146.33 €
4 L
Please to purchase this item. Need a web account? Register with us today!
11337328
SDS View Documents
Thermo Scientific Alfa Aesar
030698.K7
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Acetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glasswares due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon, photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336-H373

missing translation for 'dotInformation' : DOT Class: 3; Packing Group: II

missing translation for 'einecsNumber' : 200-662-2

missing translation for 'rtecsNumber' : AL3150000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, ACS, 99.5+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.