missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™

ACS reagent,≥99.5%

37.39 € - 763.30 €

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15671600
SDS View Documents
Honeywell-Riedel-de Haen
179124-500ML
500 mL Glass Bottle N/A N/A N/A
15651600
SDS View Documents
Honeywell-Riedel-de Haen
179124-2L
2 L Glass Bottle N/A N/A N/A
15631600
SDS View Documents
Honeywell-Riedel-de Haen
179124-2.5L
2.5 L Glass Bottle
179.70 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15681600
SDS View Documents
Honeywell-Riedel-de Haen
179124-6X1L
6 x 1 L Glass Bottle
401.89 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15661600
SDS View Documents
Honeywell-Riedel-de Haen
179124-4X2.5L
4 x 2.5 L Glass Bottle
455.79 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15641600
SDS View Documents
Honeywell-Riedel-de Haen
179124-20L
20 L Stainless Steel Can N/A N/A N/A
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.