Learn More
Fisherbrand™ Acid Neutralizing Chemical Specialty Spill Kit
Spill kit containing chemical neutralizing polymers, which provide safe spill control.
Brand:  Fisherbrand™ FBSKABKTACIDE
Product Code. 16543852
  • 302.00 € / 1 set
Explore more special offers
Description
Description
  • Contains chemical neutralizing polymers
  • Includes color change indicator that reflects neutralization
Specifications
Specifications
Chemical Sorbent
2 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x HAZ412 (813850), 10 x UN100-E (813842), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS (198790), 1 x SPC-ACID, 1 x Bucket, 1 x Scoop and Scraper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Acid Neutralizing Chemical Specialty Spill Kit > Quantity: 1/Ea.