Learn More
Micronova™ ACME Thread Adapters
The two versions of stainless steel ACME adapters connect threaded brush and mop adapters to Quick Disconnect, SSU Series Handles (SSA-3), or connect Quick Disconnect mop adapters to threaded handles (SSA-2).
Brand:  Micronova™ SSA-3
 View more versions of this product
Product Code. 12857033
  • 60.65 € / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Micronova ACME Thread Adapters serve as the connection between various Micronova Adapters and either Threaded or Quick Disconnect handles. The SSA-2 serves to connect a range of Quick Disconnect mop adapters and cleaning tools with threaded handles, such as STA Series Handles. The SSA-3 serves to connect a range of threaded brushes and mop adapters, such as QPSL series, with the Quick Disconnect stainless steel SSU Series Handles.
  • ACME threaded adapters
  • Stainless steel with polyketone connectors
  • Connects various Adapters to either Threaded or Quick Disconnect handles
  • SSA-2 connects threaded handles to QD mop and tool adapters
  • SSA-3 connects threaded mop and brush adapters to QD SSU Handles
  • Autoclavable
Order Info
Attach to a stainless-steel handle (sold separately) with 19-035-362B Thread Adapter.
Specifications
Specifications
Male Thread Adapter
Non-sterile
Autoclavable
Floors, Walls, Isolators
5.75 in.
14.6 cm
Threaded Connector for Handles and QD Mop Adapters and Tools
Electropolished Stainless Steel (304), Polyketone