missing translation for 'onlineSavingsMsg'
Learn More
Ansell ActivArmr™ 43-217 Grain Goatskin Leather TIG Welding Gloves Product Code.: 16815230
In Stock
to see stock.
Add to basket

Ansell ActivArmr™ 43-217 Grain Goatskin Leather TIG Welding Gloves

Product Code. 16815230 Shop All Ansell Products
11
Click to view available options
Size:
9
10
11
This item is not returnable. View return policy

Product Code. 16815230

Brand: Ansell 43217110

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Premium full grain goatskin leather combined with para-aramid welted seams ensure durability in use while also providing protection from heat, flames, sparks, and weld spatter.

  • Premium full grain goatskin leather and welted seams with para-aramid thread ensure durability in use
  • 3-dimensional thumb design greatly increases fine dexterity for precise welding control
  • Long split cowhide cuff extends protection to the forearms
  • Provides protection from heat, flames, sparks and weld spatter
  • Suitable for TIG welding as well as grinding, brazing, metal fabrication, and maintenance
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Cuff Material Split Cowhide Leather
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A1
Material Grain Goatskin Leather
Size 11
For Use With (Application) Automotive, Machinery, and Equipment
Glove Cuff Color Yellow
Glove Cuff Style Gauntlet
Series ActivArmr™ 43-217
Length (English) 14.2 in.
Length (Metric) 36 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell ActivArmr™ 43-217 Grain Goatskin Leather TIG Welding Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.