Learn More
Honeywell Burdick & Jackson™ Activator Reagent, 0.5M ETT in Acetonitrile
BioSyn™, for DNA/RNA synthesis
Brand:  Honeywell Burdick & Jackson™ BR725-2.5GL
 View more versions of this product
Product Code. 17195451
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
75-05-8
0.809 g/cm3
GLASS BOTTLE, GL45
4 x 2.5 L