Learn More
Honeywell Burdick & Jackson™ Activator Reagent, 0.5M ETT in Acetonitrile
BioSyn™, for DNA/RNA synthesis
3615.00 €
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging Type | Price | Quantity & Availability | ||||
17195451
|
Honeywell Burdick & Jackson™
BR725-2.5GL |
4 x 2.5 L | GLASS BOTTLE, GL45 | N/A | |||||
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
75-05-8 | |
0.809 g/cm3 |