Learn More
Socorex™ Acura™ Manual 825 Micropipette Triopack
Classic and universal, these micropipettes are intended for all applications and come in a select three-pack. Socorex™ Acura™ Manual 825 Micropipette Triopack micropipettes are designed for comfort and sturdiness.
Brand:  Socorex™ 825.TRIO.S
 View more versions of this product
Product Code. 10690833
  • 856.00 € / 1 set
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ergonomic shape.
  • Lightweight.
  • Uultra-soft plunger stroke.
  • Smart and reliable volume adjustment:
    • State-of-the-art micrometric setting is performed smoothly and precisely by turning push button.
    • Finely machined click-stops and free rotating cap prevent any unwanted volume alteration.
  • Digital display of set volume on the front of the instrument is easily visible during pipetting.
  • Autoclavable.
Specifications
Specifications
EN ISO 8655, DIN 12650, GMP, GLP, GBEA, NCCLS and CE IVD Directive 98/79 EEC on medical devices
Acura
2 to 20, 20 to 200, 100 to 1000 μL
3/Pk.
Autoclavable
Micropipette Triopack
3 Acura™ micropipettes, manual, spike samples, Qualitips™ 20/200/1000μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products