missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adapt-a-Rack™

Accommodates tubes 12 to 30 mm dia. (5 to 50 mL). Adapt-a-Rack™ holds tubes in a firm, upright position.

Brand:  Fisherbrand™ HS120188

 View more versions of this product

Product Code. 15350370

  • 16.45 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Made from Polyoxymethylene (POM).
  • Racks can be linked together for additional holding capacity (Linked racks should be supported underneath when carrying).
  • Open sides allow for label viewing.
  • Flat base to hold tubes upright.
  • Available in solid or a combination of colors for easy coding identification.
  • Autoclavable.
Specifications

Specifications

Multi Stand Tube Rack
White/White
POM
Rectangular, Rounded Corners
Yes, Interlocking
2 Pk.
Autoclavable
Tubes (5 to 50 mL)
4
None
Non-sterile
12 to 30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.