Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100245
 View more versions of this product
Product Code. 10302711
  • 123.00 € / Pack of 10
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cones
  • Acc. to DIN 12249 form B
  • Standard, conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
NS 45/40
2.3 mm
40 mm
Duran Glass