Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
29.00 € - 123.00 €
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10040331
|
Lenz Laborglasintrumente
01100212 |
NS 12/21 |
45.00 €
Pack of 10 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
10343281
|
Lenz Laborglasintrumente
01100214 |
NS 14/23 |
29.00 €
Pack of 10 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
10792512
|
Lenz Laborglasintrumente
01100219 |
NS 19/26 |
31.00 €
Pack of 10 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
10702522
|
Lenz Laborglasintrumente
01100224 |
NS 25/29 |
39.00 €
Pack of 10 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
10303331
|
Lenz Laborglasintrumente
01100229 |
NS 29/32 |
40.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10302711
|
Lenz Laborglasintrumente
01100245 |
NS 45/40 |
123.00 €
Pack of 10 |
Estimated Shipment: 06-03-2025
Log in to see stock. |
|||||
Description
- Cones
- Acc. to DIN 12249 form B
- Standard, conical ground cones
- Made of DURAN™ tubing
Specifications
Duran Glass |