Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100229
 View more versions of this product
Product Code. 10303331
  • 40.00 € / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cones
  • Acc. to DIN 12249 form B
  • Standard, conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
NS 29/32
2 mm
26 mm
Duran Glass