missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters For Round Bucket in Rotor A-4-38
Wide variety of adapters for all blood collection tubes and cell culture tube types
Brand: Eppendorf™ 5702718009
107.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adapter for Rotor A-4-38
- For centrifugation of blood collection tubes or conical tubes
Specifications
Adapter | |
85 mL | |
38 mm | |
2 Pack |
Oak Ridge | |
Round bucket in Rotor A-4-38 | |
4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction