missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adefovir dipivoxil
83.82 € - 219.09 €
Specifications
Format | Powder |
---|---|
Molecular Weight (g/mol) | 501.47 |
Product Type | Adefovir dipivoxil |
Source | synthetic source |
Content And Storage | Freezer -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16651822
|
Thermo Scientific Chemicals
463080010 |
1 g |
83.82 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16661822
|
Thermo Scientific Chemicals
463080100 |
10 g |
219.09 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Powder | |
Adefovir dipivoxil | |
Freezer -20°C | |
normal conditions |
501.47 | |
synthetic source | |
98.0 to 102.0 % (Titration with HClO4) | |
White to almost white |
Safety and Handling
missing translation for 'shelfLife' : 5 years
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Adefovir dipivoxil
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Adefovir dipivoxil
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title