missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Adenine, 99% Product Code.: 10716181

Thermo Scientific Chemicals Adenine, 99%

Product Code. 10716181
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10716181

Brand: Thermo Scientific Chemicals 147440250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purineShow MoreShow Less
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Absorbance 0.74 to 0.78 (A250/A260), 0.36 to 0.4 (A280/A260)
CAS Min % 99.5
CAS Max % 100.0
Melting Point 360°C to 365°C
Color Colorless to Yellow
Loss on Drying 0.5% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 26, 420
Merck Index 15, 140
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in acids and alkalies,slightly soluble in ethanol,insoluble in chloroform and ether,25g/l in boiling water
Sublimation Point 220°C
Spectrophotometry 98.0 to 102.0% (lambda max. 262nm, pH 1), molar absorptivity 13100 +/- 2%, on dry basis
Formula Weight 135.13
Percent Purity 99%
Physical Form Liquid or Solid
Chemical Name or Material Adenine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466909 Certificate TypeCertificate of Analysis Date21/10/2024 Catalog Number147445000,147440250,147441000
Lot NumberA0464623 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog Number147445000,147440250,147441000
Lot NumberA0460909 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number147445000,147440250,147441000
Lot NumberA0453827 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number147445000,147440250,147441000
Lot NumberA0415074 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number147445000,147440250,147441000
compliance-icons
Product Identifier
  • Adenine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Adenine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.