missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Adenine, 99% Product Code.: 11402717

Thermo Scientific Chemicals Adenine, 99%

Product Code. 11402717
25 g
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11402717

Brand: Thermo Scientific Chemicals A14906.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purineShow MoreShow Less
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Assay Percent Range 99%
Quantity 25 g
UN Number UN2811
Beilstein 5777
Merck Index 14,152
Formula Weight 135.13
Percent Purity 99%
Chemical Name or Material Adenine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10208013 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14906.18,A14906.14,A14906.36,A14906.30
Lot NumberF4612A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14906.18,A14906.14,A14906.36,A14906.30
Lot Number10141555 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14906.18,A14906.14,A14906.36,A14906.30
Lot NumberF4612B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14906.18,A14906.14,A14906.36,A14906.30
Lot Number10037151 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14906.18,A14906.14,A14906.36,A14906.30
compliance-icons
Product Identifier
  • Adenine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Adenine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Adenine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.