Learn More
Color Coded High Resolution pH Buffer, Reagecon™
BIOPHARMA
Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.
Brand:  Reagecon H70525C
 View more versions of this product
Product Code. 11404609
  • 72.90 € / 500mL
Explore more special offers
Description
Description
  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Specifications
Specifications
0°C
7
500 mL
Yellow