missing translation for 'onlineSavingsMsg'
Learn More

Color Coded High Resolution pH Buffer, Reagecon™
BIOPHARMA

Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.

32.35 € - 72.90 €

Specifications

Melting Point 0°C
Physical Form Liquid
Quantity 500 mL
View More Specs

Products 6
Product Code Brand Quantity pH Price Quantity & Availability  
Product Code Brand Quantity pH Price Quantity & Availability  
11778465
BIOPHARMA
View Documents Promotion Details
Reagecon
110000C
500 mL 10.00
37.85 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11494599
BIOPHARMA
View Documents Promotion Details
Reagecon
H40525C
500 mL 4
62.75 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11758465
BIOPHARMA
View Documents Promotion Details
Reagecon
104000C
500 mL 4.00
42.45 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11404609
BIOPHARMA
View Documents Promotion Details
Reagecon
H70525C
500 mL 7
72.90 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15214643
BIOPHARMA
View Documents Promotion Details
Reagecon
H70525C
500 mL 7
32.35 €
Each
Please to purchase this item. Need a web account? Register with us today!
11768465
BIOPHARMA
View Documents Promotion Details
Reagecon
107000C
500 mL 7.00
42.45 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Specifications

Specifications

0°C
Liquid
500 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BUFFER SOLUTION pH 10.000 ± 0.010 @ 20°C

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : Boric Acid, DisodiumSalt

SDS
Documents

Documents

Product Certifications