missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats
Reduce contamination in high traffic areas. ITW Chemische CleanStep™ Adhesive Cleanroom Mats are constructed with either 30 or 60 layers. Each removable sheet has a consistent adhesive coating designed to trap particles and prevent transfer.
Brand: ITW Chemische Produkte™ AMA184682W
210.90 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
This item is not returnable.
View return policy
Description
- Designed to capture particulates from foot and wheel borne traffic prior to entry into cleanrooms or controlled environments
- Provide a portable contamination control solution
- Can be used with or without frames (sold separately)
- Contain anti-microbial preservatives that prevent microbial growth
- Consistent adhesive coating on each layer of 30 and 60 layer mats
- Possible applications include cleanroom and critical environment entry points, gowning rooms or anterooms in the semiconductor, biotechnology, medical device, pharmaceutical processing, aerospace, and automotive markets
Specifications
18 x 46 in. (46 x 117 cm) | |
60 layer | |
White | |
Polyethylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats > Size: 18 x 46 in. (46 x 117cm); 60 layers; White
Spot an opportunity for improvement?Share a Content Correction