Learn More
Grant Instruments™ Advanced Air Cleaner Recirculator, 2 Lamps
Powerful/efficient UV cleaner-recirculator. Two UV lamps providing air disinfection in hospitals, research laboratories and clinics. The UV air flow cleaner-recirculator consists of 2 germicidal UV lamps, a fan, unit equipped with dust filters and a control unit, confined in a flow through chamber.
Brand:  Grant Instruments™ UVR-Mi UK
Product Code. 17775149
  • 872.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The inner mirror surface of the recirculator chamber reflects ultraviolet rays thereby increasing the density of UV radiation and enhancing the disinfection effect.
  • Two UV-lamps
  • Full user protection from direct UV radiation;
  • Two operation modes — short time (under timer control) and continuous operation
  • Digital control unit allows tracking of overall UV lamp operating time
  • 1 min-24 hours/non stop timer
Recirculators fixation:
  • Convenient fixation on walls (standard)
  • Mounting on a movable tripod (optional)
Specifications
Specifications
18 mW/cm2/sec, 14 m3/hour
2 Lamps
UVR-S
UV-Air Flow Cleaner-Recirculator