Learn More
AG 205
Progesterone receptor membrane component 1 (Pgrmc1) ligand
Brand:  Tocris Bioscience 6242/5
115.10 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18014312
  • 143.00 € / 5mg
Explore more special offers
Description
Description
AG 205 is a progesterone receptor membrane component 1 (Pgrmc1) ligand. Inhibits cell cycle progression and cell viability in cancer cell lines. Inhibits progesterone induced cytosolic Ca2+ increases in GnRH neurons in vitro.
Specifications
Specifications
cis-2-[[1-(4-Chlorophenyl)-1H-tetrazol-5-yl]thio]-1-(1,2,3,4,4a,9b-hexahydro-2,8-dimethyl-5H-pyrido[4,3-b]indol-5-yl)-ethanone
>98%
C22H23ClN6OS
442656-02-2
5 mg
Observed MW: 454.98