missing translation for 'onlineSavingsMsg'
Learn More

AG 205

Progesterone receptor membrane component 1 (Pgrmc1) ligand

143.00 € - 497.00 €

Specifications

Chemical Name or Material cis-2-[[1-(4-Chlorophenyl)-1H-tetrazol-5-yl]thio]-1-(1,2,3,4,4a,9b-hexahydro-2,8-dimethyl-5H-pyrido[4,3-b]indol-5-yl)-ethanone
CAS 442656-02-2
Assay Percent Range >98%
Molecular Formula C22H23ClN6OS
Formula Weight Observed MW: 454.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18707926
View Documents
Tocris Bioscience
6242/25
25 mg
497.00 €
25mg
Please to purchase this item. Need a web account? Register with us today!
18014312
View Documents
Tocris Bioscience
6242/5
5 mg
143.00 €
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AG 205 is a progesterone receptor membrane component 1 (Pgrmc1) ligand. Inhibits cell cycle progression and cell viability in cancer cell lines. Inhibits progesterone induced cytosolic Ca2+ increases in GnRH neurons in vitro.
Specifications

Specifications

cis-2-[[1-(4-Chlorophenyl)-1H-tetrazol-5-yl]thio]-1-(1,2,3,4,4a,9b-hexahydro-2,8-dimethyl-5H-pyrido[4,3-b]indol-5-yl)-ethanone
>98%
Observed MW: 454.98
442656-02-2
C22H23ClN6OS
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications