missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Agarose
Molecular Biology Grade Agarose.
Brand: Melford A20090-100.0
This item is not returnable.
View return policy
Description
Molecular Biology Grade Agarose.
Molecular Biology Grade Agarose. High Purity for preparative and analytical electrophoresis of nucleic acids ≥1000bp. Forms a solid gel at low concentration. Product is RNase/DNase free.
Gel Strength, 1%: ≥1200 g/cm2
Gel Strength, 1.5%: ≥2700 g/cm2
Gel Point: 35°C to 37°C
Melting Point: 87°C to 89°C
EEO: 0.09 to 0.12
Sulfate: ≥0.03%
Loss on Drying: <10%
DNA Binding: None Detected
Specifications
Agarose | |
9012-36-6 | |
100 g | |
Molecular Biology |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction