Learn More
Melford Agarose
Molecular Biology Grade Agarose.
143.00 € - 1260.00 €
Specifications
Chemical Name or Material | Agarose |
---|---|
CAS | 9012-36-6 |
Grade | Molecular Biology |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30017982
|
Melford
A20090-1000.0 |
1 kg |
1260.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017981
|
Melford
A20090-100.0 |
100 g |
243.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017983
|
Melford
A20090-50.0 |
50 g |
143.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017984
|
Melford
A20090-500.0 |
500 g |
687.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molecular Biology Grade Agarose.
Molecular Biology Grade Agarose. High Purity for preparative and analytical electrophoresis of nucleic acids ≥1000bp. Forms a solid gel at low concentration. Product is RNase/DNase free.
Gel Strength, 1%: ≥1200 g/cm2
Gel Strength, 1.5%: ≥2700 g/cm2
Gel Point: 35°C to 37°C
Melting Point: 87°C to 89°C
EEO: 0.09 to 0.12
Sulfate: ≥0.03%
Loss on Drying: <10%
DNA Binding: None Detected
Specifications
Agarose | |
9012-36-6 | |
Molecular Biology |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.