Learn More
Melford Agarose Low EEO
Molecular Biology Grade.
Brand: Melford A20080-1000.0
This item is not returnable.
View return policy
Description
Molecular Biology Grade.
Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.Gelling Temperature:
34.5 -37.5°C
Sulfate:
0.15
EEO:
0.09- 0.13
DNA Binding:
None Detected
DNA and RNase:
None Detected
Specifications
Agarose, Low EEO | |
9012-36-6 | |
1 kg | |
Molecular Biology |