missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Agarose Low EEO
Molecular Biology Grade.
137.00 € - 1905.00 €
Specifications
Chemical Name or Material | Agarose, Low EEO |
---|---|
CAS | 9012-36-6 |
Grade | Molecular Biology |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30017977
|
Melford
A20080-50.0 |
50 g |
137.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017975
|
Melford
A20080-100.0 |
100 g |
263.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017976
|
Melford
A20080-1000.0 |
1 kg |
1905.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017978
|
Melford
A20080-500.0 |
500 g |
959.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molecular Biology Grade.
Molecular Biology Grade. High purity for preparative and analytical electrophoresis of nucleic acids >1000 bp.Gelling Temperature:
34.5 -37.5°C
Sulfate:
0.15
EEO:
0.09- 0.13
DNA Binding:
None Detected
DNA and RNase:
None Detected
Specifications
Agarose, Low EEO | |
9012-36-6 | |
Molecular Biology |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title