Learn More
Syngene™ Airclean™ Operator Protection Microscope Sash
GREENER_CHOICE
Operator Protection Enclosure fitted with AirSafe™ Automatic Safety Controller
Brand:  Syngene™ SYN-32OPAS-E
 View more versions of this product
Product Code. 16522972
  • 8830.00 € / Each
Explore more special offers
Description
Description
  • Ductless microscope enclosures provide an ISO 5 work environment for sample preparation
  • Downflow microscope enclosure provides a clean environment with UV light for irradiation between processes
  • UVTect™ microprocessor controller provides one-touch operation of all enclosure features
  • Each enclosure is designed to work with a wide range of microscopes
  • Access port provides easy access to the eye piece and viewing platform
  • Configuration does not provide operator protection
Specifications
Specifications
Laminar Flow
101.6 cm
81.28 cm
61 cm
32 in.