missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Airtight Box

Store a wide variety of substances in this versatile plastic box. Azlon™ Airtight Box is lightweight and unbreakable in normal use.

Brand:  Azlon BWY036

22.56 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12059632

  • 24.35 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from tough HDPE
  • Tight fitting snap on lids create an airtight seal
  • Suitable for food contact applications in accordance with EU regulations
  • Suitable for general and refrigerated storage down to -50°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Airtight Box
Snap-on lid
90 mm
100 mm
HDPE
For general and refrigerated storage down to –50°C
210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers