missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Gray
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
30.00 € - 70.00 €
Specifications
Color | Gray |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | |||||
17742068
|
Notrax
410S0323GY |
91 cm | 60 cm |
30.00 €
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
17772068
|
Notrax
410S0335GY |
150 cm | 91 cm |
70.00 €
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
|||||
17752058
|
Notrax
410C0324GY |
1830 cm | 60 cm | N/A | ||||||
17782058
|
Notrax
410C0336GY |
1830 cm | 91 cm | N/A | ||||||
17712068
|
Notrax
410C0348GY |
1830 cm | 122 cm | N/A | ||||||
Description
- Ribbed pattern enhances traction
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Gray | |
Microcellular Vinyl | |
Industrial and Commercial Matting |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Gray