Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Gray
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
Brand: Notrax 410C0324GY
This item is not returnable.
View return policy
Description
- Ribbed pattern enhances traction
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Gray | |
1830 cm | |
Industrial and Commercial Matting |
60 ft. | |
Microcellular Vinyl | |
60 cm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Gray > Matting; Length: 1830 cm; Width: 60 cm
Spot an opportunity for improvement?Share a Content Correction