Learn More
Honeywell™ Airvisor Carbon Cartridge
Carbon filter suitable for all Airvisor air respirators.
Brand:  Honeywell™ 1001672
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12662663
  • 68.00 € / Each
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for all Airvisor models
  • Removes unpleasant odor
  • Should be replaced by 1000 hours or when unpleasant smell is experienced
  • Comes with two replacement O-rings
  • Part of an air respirator system certified to EN14594
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Removing Odor
Airvisor Air Respirators
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Honeywell™ Airvisor Carbon Cartridge > For use with Airvisor air respirators