Learn More
Fisherbrand™ Polyethylene Sample Bags
Description
- Secure, contaminant free bags to ensure dependable analysis results
- Made from heavy gauge, virgin polyethylene that meets FDA specifications
- Can accommodate solid, semi solid and liquid materials
- Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
- Economical and efficient way to collect, contain and carry samples
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Product Title
Fisherbrand™ Polyethylene Sample Bags > Capacity: 1.26L; Closure Type: Flat Wire Closure
Spot an opportunity for improvement?Share a Content Correction