Learn More
BINDER™ Alarm Output
Alarm output, zero-voltage, accessible via 3-pin clamping sleeve
Brand:  BINDER™ 8012-2502
 View more versions of this product
Product Code. 30238373
  • 443.00 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Alarm output
  • Zero-voltage
  • Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
  • Acoustic signal that can be switched off
Specifications
Specifications
KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600
Copper
Analog Output
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Alarm Output > For use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600