missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AlbuMAX™ I Lipid-Rich BSA
AlbuMAX™ I Lipid-Rich BSA
129.00 € - 3225.00 €
Specifications
Content And Storage | Store in refrigerator (2–8°C). |
---|---|
Product Type | BSA |
Product Line | AlbuMAX™, Gibco™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11530376
|
Gibco™
11020021 |
25 g |
129.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10114672
|
Gibco™
11020039 |
1 kg |
3225.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 0.1%.Origin:
New Zealand
Specifications
Store in refrigerator (2–8°C). | |
BSA | |
AlbuMAX™, Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ AlbuMAX™ I Lipid-Rich BSA